Product Details
- SNP ID
-
rs151104269
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:99554313 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCCCGTGGGGTGTGCGCTCACCT[C/T]GGTTTTCCTTGGCCAGGTTGTCGGC
- Phenotype
-
MIM: 601464
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AFF3
PubMed Links
Gene Details
- Gene
- AFF3
- Gene Name
- AF4/FMR2 family member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001025108.1 |
3728 |
Missense Mutation |
CAA,CGA |
Q1211R |
NP_001020279.1 |
NM_002285.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q1186R |
NP_002276.2 |
XM_005263943.3 |
3728 |
Missense Mutation |
CAA,CGA |
Q1186R |
XP_005264000.2 |
XM_005263945.3 |
3728 |
Missense Mutation |
CAA,CGA |
Q855R |
XP_005264002.1 |
XM_011511169.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q1210R |
XP_011509471.1 |
XM_011511170.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q1263R |
XP_011509472.1 |
XM_011511171.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q1195R |
XP_011509473.1 |
XM_011511172.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q1195R |
XP_011509474.1 |
XM_011511173.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q1211R |
XP_011509475.1 |
XM_011511174.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q1211R |
XP_011509476.1 |
XM_011511175.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q1186R |
XP_011509477.1 |
XM_011511176.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q1186R |
XP_011509478.1 |
XM_011511177.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q1186R |
XP_011509479.1 |
XM_011511178.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q825R |
XP_011509480.1 |
XM_011511179.2 |
3728 |
Missense Mutation |
CAA,CGA |
Q824R |
XP_011509481.1 |
XM_017004085.1 |
3728 |
Missense Mutation |
CAA,CGA |
Q854R |
XP_016859574.1 |
XM_017004086.1 |
3728 |
Missense Mutation |
CAA,CGA |
Q822R |
XP_016859575.1 |
XM_017004087.1 |
3728 |
Missense Mutation |
CAA,CGA |
Q821R |
XP_016859576.1 |
View Full Product Details