Product Details

SNP ID
rs151206448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3498966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTAGTTCCCAGGCAGCACTGCTA[C/G]GCGGTCTGTGCCAGAAATTTCACGT
Phenotype
MIM: 613400
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADI1 PubMed Links

Gene Details

Gene
ADI1
Gene Name
acireductone dioxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306077.1 3357 Silent Mutation GCC,GCG A173A NP_001293006.1
NM_018269.3 3357 Silent Mutation GCC,GCG A179A NP_060739.2

View Full Product Details