Product Details

SNP ID
rs151290758
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32225870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTACCTCACCCAACTTTGGAGG[A/G]TTAATTTGTTGTATATTAAACTTGT
Phenotype
MIM: 606831 MIM: 611148
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NLRC4 PubMed Links

Gene Details

Gene
NLRC4
Gene Name
NLR family CARD domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199138.1 Intron NP_001186067.1
NM_001199139.1 Intron NP_001186068.1
NM_001302504.1 Intron NP_001289433.1
NM_021209.4 Intron NP_067032.3
XM_017004619.1 Intron XP_016860108.1
Gene
SLC30A6
Gene Name
solute carrier family 30 member 6
There are no transcripts associated with this gene.

View Full Product Details