Product Details

SNP ID
rs121918244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121781772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTCTGACATAGTCATCCACTC[G/A]TTTCTTCAGTTCAACTCGGCGTGCA
Phenotype
MIM: 609237
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
IQCB1 PubMed Links

Gene Details

Gene
IQCB1
Gene Name
IQ motif containing B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023570.3 1280 Intron NP_001018864.2
NM_001023571.3 1280 Intron NP_001018865.2
NM_001319107.1 1280 Intron NP_001306036.1
XM_005247911.3 1280 Intron XP_005247968.1
XM_005247912.2 1280 Intron XP_005247969.1
XM_011513335.2 1280 Intron XP_011511637.1
XM_017007537.1 1280 Intron XP_016863026.1
XM_017007538.1 1280 Nonsense Mutation CGA,TGA R277* XP_016863027.1
XM_017007539.1 1280 Intron XP_016863028.1

View Full Product Details