Product Details

SNP ID
rs138258387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:190428665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGTCAAACTAAATTTTAGATAT[C/T]TATTGATGTTTATTGTTTTGTTCAA
Phenotype
MIM: 614786
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM207 PubMed Links

Gene Details

Gene
TMEM207
Gene Name
transmembrane protein 207
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207316.2 1438 UTR 3 NP_997199.1

View Full Product Details