Product Details

SNP ID
rs138461976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:141232975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGAATGGGGAAGTGTAGGGGG[A/G]TGGATGGGGGATCGGAGAGTGCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PXYLP1 PubMed Links

Gene Details

Gene
PXYLP1
Gene Name
2-phosphoxylose phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037172.2 Intron NP_001032249.1
NM_001282728.1 Intron NP_001269657.1
NM_152282.4 Intron NP_689495.1
XM_005247903.3 Intron XP_005247960.1
XM_011513314.2 Intron XP_011511616.1
XM_017007510.1 Intron XP_016862999.1
XM_017007511.1 Intron XP_016863000.1

View Full Product Details