Product Details

SNP ID
rs138629895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149320490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGTCATTTTCTCTTGCCCATTG[A/G]GCATGTTCCCTAGCTGCTCACTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TM4SF18 PubMed Links

Gene Details

Gene
TM4SF18
Gene Name
transmembrane 4 L six family member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184723.1 1684 UTR 3 NP_001171652.1
NM_138786.3 1684 UTR 3 NP_620141.1

View Full Product Details