Product Details

SNP ID
rs139146415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:140678509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAACTGCTACTACTATGAGAGC[C/T]GCTGCTGCCGCAATACCATCATCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM42 PubMed Links

Gene Details

Gene
TRIM42
Gene Name
tripartite motif containing 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152616.4 486 Missense Mutation CGC,TGC R94C NP_689829.3
XM_011512740.2 486 Missense Mutation CGC,TGC R94C XP_011511042.1

View Full Product Details