Product Details

SNP ID
rs139284369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:99818016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGGAGACGAGTGGTGGGAGAAC[C/G]AGCCGACTGGAGCAGGCAGCAGCCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CMSS1 PubMed Links

Gene Details

Gene
CMSS1
Gene Name
cms1 ribosomal small subunit homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167924.1 183 Intron NP_001161396.1
NM_032359.3 183 Missense Mutation CAG,GAG Q13E NP_115735.2
Gene
HP09053
Gene Name
uncharacterized LOC101929357
There are no transcripts associated with this gene.

View Full Product Details