Product Details

SNP ID
rs139579015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151880615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGAGGCTGCCCTGGAAAAGTA[C/T]TACCTTTCCATTTTTTATGGGATTG
Phenotype
MIM: 606381
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AADACL2-AS1 PubMed Links

Gene Details

Gene
AADACL2-AS1
Gene Name
AADACL2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SUCNR1
Gene Name
succinate receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033050.5 177 Silent Mutation TAC,TAT Y24Y NP_149039.2

View Full Product Details