Product Details

SNP ID
rs139669117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151198562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATAGGATCAAAGCACAGGTTCGA[C/T]ACAGCCAGGAGCAGTGTAGCCTCTT
Phenotype
MIM: 611318
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR171 PubMed Links

Gene Details

Gene
GPR171
Gene Name
G protein-coupled receptor 171
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013308.3 1065 Silent Mutation GTA,GTG V275V NP_037440.3
XM_005247402.3 1065 Silent Mutation GTA,GTG V275V XP_005247459.1
XM_005247403.3 1065 Silent Mutation GTA,GTG V275V XP_005247460.1
XM_017006274.1 1065 Silent Mutation GTA,GTG V275V XP_016861763.1
Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 1065 Intron NP_443728.3
XM_006713487.3 1065 Intron XP_006713550.1
XM_011512390.2 1065 Intron XP_011510692.1
XM_011512394.2 1065 Intron XP_011510696.1
XM_011512399.2 1065 Intron XP_011510701.1
XM_017005676.1 1065 Intron XP_016861165.1
XM_017005677.1 1065 Intron XP_016861166.1
XM_017005678.1 1065 Intron XP_016861167.1
XM_017005679.1 1065 Intron XP_016861168.1
XM_017005680.1 1065 Intron XP_016861169.1
XM_017005681.1 1065 Intron XP_016861170.1

View Full Product Details