Product Details

SNP ID
rs139730198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32996726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCACATTAAAAACAGTGACAT[C/T]ATCATTTGAGTACAAATTTTATTTA
Phenotype
MIM: 611458
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLB1 PubMed Links

Gene Details

Gene
GLB1
Gene Name
galactosidase beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000404.3 2392 UTR 3 NP_000395.2
NM_001079811.2 2392 UTR 3 NP_001073279.1
NM_001135602.2 2392 UTR 3 NP_001129074.1
NM_001317040.1 2392 UTR 3 NP_001303969.1

View Full Product Details