Product Details

SNP ID
rs140085841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98391146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCTGCATCCCATGATTGAAGTA[C/G]AGTTTCTGTTGAGGTTAACTTTCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5K3 PubMed Links

Gene Details

Gene
OR5K3
Gene Name
olfactory receptor family 5 subfamily K member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005516.1 481 Missense Mutation CAG,GAG Q161E NP_001005516.1

View Full Product Details