Product Details
- SNP ID
-
rs140250886
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:146194412 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTTGATGGCTTGCTGTGTCTCTC[C/T]ATCTTGAACGTTGTGGTGGAGATCT
- Phenotype
-
MIM: 607612
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLSCR4
PubMed Links
Gene Details
- Gene
- PLSCR4
- Gene Name
- phospholipid scramblase 4
There are no transcripts associated with this gene.
View Full Product Details