Product Details

SNP ID
rs140262971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23893116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTAGTTCTCAGAATTAGAGTT[C/T]CCTTTGTTTTTCCTCCCAGGCAAAG
Phenotype
MIM: 604496 MIM: 602916
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NKIRAS1 PubMed Links

Gene Details

Gene
NKIRAS1
Gene Name
NFKB inhibitor interacting Ras like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020345.3 799 Silent Mutation GGA,GGG G186G NP_065078.1
XM_005265076.3 799 Silent Mutation GGA,GGG G186G XP_005265133.1
XM_005265077.4 799 Silent Mutation GGA,GGG G186G XP_005265134.1
XM_005265078.3 799 Silent Mutation GGA,GGG G186G XP_005265135.1
Gene
UBE2E1
Gene Name
ubiquitin conjugating enzyme E2 E1
There are no transcripts associated with this gene.

View Full Product Details