Product Details

SNP ID
rs140978440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169793643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTACAGCTACTTTTTCATGGCT[A/G]TTGACCTACTGGAACAAGAGCAAAA
Phenotype
MIM: 606042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC34 PubMed Links

Gene Details

Gene
LRRC34
Gene Name
leucine rich repeat containing 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172779.1 1355 Nonsense Mutation CAG,TAG Q463* NP_001166250.1
NM_001172780.1 1355 Intron NP_001166251.1
NM_153353.4 1355 Nonsense Mutation CAG,TAG Q431* NP_699184.2
XM_006713508.3 1355 Nonsense Mutation CAG,TAG Q445* XP_006713571.1
XM_011512442.2 1355 Nonsense Mutation CAG,TAG Q462* XP_011510744.1
XM_017005746.1 1355 Nonsense Mutation CAG,TAG Q401* XP_016861235.1
Gene
MYNN
Gene Name
myoneurin
There are no transcripts associated with this gene.

View Full Product Details