Product Details

SNP ID
rs141226859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113288351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAACTTCATGTGGAAGTGCAGCA[C/T]GGTTTTCAGGAAAGCAGTATTTTAA
Phenotype
MIM: 608708
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BOC PubMed Links

Gene Details

Gene
BOC
Gene Name
BOC cell adhesion associated, oncogene regulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301861.1 8266 Intron NP_001288790.1
NM_001301867.1 8266 Intron NP_001288796.1
NM_033254.3 8266 Intron NP_150279.1
XM_005247891.2 8266 Intron XP_005247948.1
XM_005247892.2 8266 Intron XP_005247949.1
XM_005247897.2 8266 Intron XP_005247954.1
XM_011513305.2 8266 Intron XP_011511607.1
XM_017007441.1 8266 Intron XP_016862930.1
XM_017007442.1 8266 Intron XP_016862931.1
XM_017007443.1 8266 Intron XP_016862932.1
XM_017007444.1 8266 Intron XP_016862933.1
XM_017007445.1 8266 Intron XP_016862934.1
XM_017007446.1 8266 Intron XP_016862935.1
XM_017007447.1 8266 Intron XP_016862936.1
XM_017007448.1 8266 Intron XP_016862937.1
XM_017007449.1 8266 Intron XP_016862938.1
XM_017007450.1 8266 Intron XP_016862939.1
XM_017007451.1 8266 Intron XP_016862940.1
XM_017007452.1 8266 Intron XP_016862941.1
XM_017007453.1 8266 Intron XP_016862942.1
XM_017007454.1 8266 Intron XP_016862943.1
XM_017007455.1 8266 Intron XP_016862944.1
Gene
CFAP44
Gene Name
cilia and flagella associated protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164496.1 8266 UTR 3 NP_001157968.1
NM_018338.3 8266 Intron NP_060808.2
XM_005247617.3 8266 UTR 3 XP_005247674.1
XM_006713696.2 8266 UTR 3 XP_006713759.1
XM_011512977.2 8266 UTR 3 XP_011511279.1
XM_011512978.2 8266 UTR 3 XP_011511280.1
XM_011512979.2 8266 UTR 3 XP_011511281.1
XM_011512980.2 8266 Intron XP_011511282.1
XM_011512981.1 8266 Intron XP_011511283.1
XM_011512982.2 8266 Intron XP_011511284.1
XM_011512983.2 8266 Intron XP_011511285.1
XM_011512984.2 8266 Intron XP_011511286.1
XM_017006842.1 8266 Intron XP_016862331.1
XM_017006843.1 8266 Intron XP_016862332.1
XM_017006844.1 8266 UTR 3 XP_016862333.1
XM_017006845.1 8266 UTR 3 XP_016862334.1
XM_017006846.1 8266 UTR 3 XP_016862335.1
XM_017006847.1 8266 UTR 3 XP_016862336.1
XM_017006848.1 8266 Intron XP_016862337.1
XM_017006849.1 8266 Intron XP_016862338.1

View Full Product Details