Product Details

SNP ID
rs141361690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9870049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTTGGCAGGCTTATGGGAGAG[C/G]GACAGTGGGTGCCTTGTCCCCTGCA
Phenotype
MIM: 612120
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CIDEC PubMed Links

Gene Details

Gene
CIDEC
Gene Name
cell death inducing DFFA like effector c
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199551.1 473 Silent Mutation TCC,TCG S139S NP_001186480.1
NM_001199552.1 473 Silent Mutation TCC,TCG S129S NP_001186481.1
NM_001199623.1 473 Silent Mutation TCC,TCG S142S NP_001186552.1
NM_001321142.1 473 Silent Mutation TCC,TCG S129S NP_001308071.1
NM_001321143.1 473 Silent Mutation TCC,TCG S55S NP_001308072.1
NM_001321144.1 473 Silent Mutation TCC,TCG S55S NP_001308073.1
NM_022094.3 473 Silent Mutation TCC,TCG S129S NP_071377.2

View Full Product Details