Product Details
- SNP ID
-
rs142030218
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:157148500 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTAAGGTGAGGAGAACCATGATTA[C/T]GATGTCTTCGAGGGCTTTCACTGTG
- Phenotype
-
MIM: 613384
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCNL1
PubMed Links
Gene Details
- Gene
- CCNL1
- Gene Name
- cyclin L1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001308185.1 |
1459 |
Intron |
|
|
NP_001295114.1 |
NM_020307.3 |
1459 |
Missense Mutation |
CAT,CGT |
H441R |
NP_064703.1 |
XM_005247648.3 |
1459 |
Missense Mutation |
CAT,CGT |
H286R |
XP_005247705.1 |
XM_006713710.3 |
1459 |
Missense Mutation |
CAT,CGT |
H408R |
XP_006713773.1 |
XM_011513011.2 |
1459 |
Missense Mutation |
CAT,CGT |
H286R |
XP_011511313.1 |
XM_017006891.1 |
1459 |
Missense Mutation |
CAT,CGT |
H235R |
XP_016862380.1 |
XM_017006892.1 |
1459 |
Missense Mutation |
CAT,CGT |
H214R |
XP_016862381.1 |
XM_017006893.1 |
1459 |
Missense Mutation |
CAT,CGT |
H214R |
XP_016862382.1 |
XM_017006894.1 |
1459 |
Missense Mutation |
CAT,CGT |
H214R |
XP_016862383.1 |
XM_017006895.1 |
1459 |
Missense Mutation |
CAT,CGT |
H214R |
XP_016862384.1 |
XM_017006896.1 |
1459 |
Missense Mutation |
CAT,CGT |
H214R |
XP_016862385.1 |
XM_017006897.1 |
1459 |
Missense Mutation |
CAT,CGT |
H202R |
XP_016862386.1 |
View Full Product Details