Product Details

SNP ID
rs142294229
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49271263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTCTCAACAATATTATGATCAT[C/T]AAAAGCCTGTTTGAGGCTGCCAGGG
Phenotype
MIM: 603486
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C3orf62 PubMed Links

Gene Details

Gene
C3orf62
Gene Name
chromosome 3 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198562.2 924 Missense Mutation AAT,GAT N241D NP_940964.1
Gene
MIR4271
Gene Name
microRNA 4271
There are no transcripts associated with this gene.

Gene
USP4
Gene Name
ubiquitin specific peptidase 4
There are no transcripts associated with this gene.

View Full Product Details