Product Details

SNP ID
rs142967558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51934668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTGTGTTGAGGAGGGCTGCCAC[C/T]GACGATATCCAGAAGGGCTGCTCCA
Phenotype
MIM: 607726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP3 PubMed Links

Gene Details

Gene
PARP3
Gene Name
poly(ADP-ribose) polymerase family member 3
There are no transcripts associated with this gene.

Gene
RRP9
Gene Name
ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004704.4 1217 Silent Mutation TCA,TCG S381S NP_004695.1

View Full Product Details