Product Details

SNP ID
rs143015321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119642486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTCCAATGATCCTTAAAGTTCA[G/T]GCGTGTGGGTTGGAGCCAAAGGGGC
Phenotype
MIM: 605823
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POPDC2 PubMed Links

Gene Details

Gene
POPDC2
Gene Name
popeye domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308333.1 1393 UTR 3 NP_001295262.1
NM_022135.3 1393 Missense Mutation CAT,CCT H362P NP_071418.2
XM_005247698.3 1393 UTR 3 XP_005247755.1

View Full Product Details