Product Details
- SNP ID
-
hCV169446003
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:50236348 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCGGACGGCGGGATGGGCTGCACC[G/T]TGAGCGCCGAGGACAAGGCGGCGGC
- Phenotype
-
MIM: 139360
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GNAI2
PubMed Links
Gene Details
- Gene
- GNAI2
- Gene Name
- G protein subunit alpha i2
There are no transcripts associated with this gene.
- Gene
- MIR5787
- Gene Name
- microRNA 5787
There are no transcripts associated with this gene.
View Full Product Details