Product Details

SNP ID
rs143915089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23947745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTCTCGTTCATTTGGCCTTAA[A/G]TTCAAAGCTGTTACAAGCTGTTACC
Phenotype
MIM: 602304
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR1D2 PubMed Links

Gene Details

Gene
NR1D2
Gene Name
nuclear receptor subfamily 1 group D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145425.1 Intron NP_001138897.1
NM_005126.4 Intron NP_005117.3
XM_006713451.3 Intron XP_006713514.1

View Full Product Details