Product Details

SNP ID
rs144093247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:167442134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAGTGAATCTAAATCTCTTCCT[G/T]TTATCTCCTGGGTGTCAGGATTTGT
Phenotype
MIM: 605587
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SERPINI2 PubMed Links

Gene Details

Gene
SERPINI2
Gene Name
serpin family I member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012303.2 1424 Missense Mutation AAA,ACA K408T NP_001012303.1
NM_006217.4 1424 Missense Mutation AAA,ACA K398T NP_006208.1

View Full Product Details