Product Details

SNP ID
rs144158073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186717549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGAGATTGTTAGATCATGAAA[C/G]TAATTACCATCCTTTTCCTCTGCTC
Phenotype
MIM: 612358
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KNG1 PubMed Links

Gene Details

Gene
KNG1
Gene Name
kininogen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000893.3 241 Missense Mutation CTA,GTA L3V NP_000884.1
NM_001102416.2 241 Missense Mutation CTA,GTA L3V NP_001095886.1
NM_001166451.1 241 Missense Mutation CTA,GTA L3V NP_001159923.1

View Full Product Details