Product Details
- SNP ID
-
rs144727670
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:152835885 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACAGCACGGTCGCCTCCACTGCC[C/G]CCGTCTCCTCGTCGTTCAAATGCGC
- Phenotype
-
MIM: 601167
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
P2RY1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1065776] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- P2RY1
- Gene Name
- purinergic receptor P2Y1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002563.4 |
1193 |
Missense Mutation |
CCC,GCC |
P35A |
NP_002554.1 |
View Full Product Details