Product Details

SNP ID
rs144743254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52706840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGGCTGAACAGTTCGGGTGGAC[G/T]GTCTATATAGTTATGGCCGGATTTG
Phenotype
MIM: 601959 MIM: 610358
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GLT8D1 PubMed Links

Gene Details

Gene
GLT8D1
Gene Name
glycosyltransferase 8 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010983.2 550 Intron NP_001010983.1
NM_001278280.1 550 Intron NP_001265209.1
NM_001278281.1 550 Intron NP_001265210.1
NM_018446.3 550 Intron NP_060916.1
NM_152932.2 550 Intron NP_690909.1
XM_017006857.1 550 Intron XP_016862346.1
XM_017006858.1 550 Intron XP_016862347.1
XM_017006859.1 550 Intron XP_016862348.1
Gene
NEK4
Gene Name
NIMA related kinase 4
There are no transcripts associated with this gene.

Gene
SPCS1
Gene Name
signal peptidase complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014041.3 550 Silent Mutation ACG,ACT T115T NP_054760.3

View Full Product Details