Product Details

SNP ID
rs144918671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194137858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGATCAATGCCATGACCTACCC[A/C]GGGCAGCCGCACCCCGCCTTGCAGG
Phenotype
MIM: 139605
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HES1 PubMed Links

Gene Details

Gene
HES1
Gene Name
hes family bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005524.3 707 Silent Mutation CCA,CCC P156P NP_005515.1

View Full Product Details