Product Details

SNP ID
rs145997815
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98087369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATGCCCATGTACTTATTCCTTG[A/G]TGGTTTAGCCTTTTCAGATGCTTGT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR5AC2 PubMed Links

Gene Details

Gene
OR5AC2
Gene Name
olfactory receptor family 5 subfamily AC member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054106.1 197 Missense Mutation GAT,GGT D66G NP_473447.1

View Full Product Details