Product Details
- SNP ID
-
rs146769270
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:147411295 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACACACACACACACACAGGGCGGA[C/T]GAGGAGGAGCTGGGTATATAGATTT
- Phenotype
-
MIM: 600470
MIM: 608948
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZIC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111509390] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZIC1
- Gene Name
- Zic family member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003412.3 |
|
Intron |
|
|
NP_003403.2 |
- Gene
- ZIC4
- Gene Name
- Zic family member 4
There are no transcripts associated with this gene.
View Full Product Details