Product Details

SNP ID
rs146905684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:16259788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCATTCTCCGCTGTAGATGCTT[C/T]ACCTTGTAGTAGTGACACAACACTG
Phenotype
MIM: 608959
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPH3 PubMed Links

Gene Details

Gene
DPH3
Gene Name
diphthamide biosynthesis 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001047434.2 1338 UTR 3 NP_001040899.1
NM_206831.2 1338 UTR 3 NP_996662.1
Gene
OXNAD1
Gene Name
oxidoreductase NAD binding domain containing 1
There are no transcripts associated with this gene.

View Full Product Details