Product Details

SNP ID
rs147057358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:142162517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGAAAAAATCAGTATGTCTATA[A/G]TATAAAAGCAATTAGTAGCACTTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GK5 PubMed Links

Gene Details

Gene
GK5
Gene Name
glycerol kinase 5 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039547.2 4847 UTR 3 NP_001034636.1
XM_011512624.2 4847 UTR 3 XP_011510926.1
XM_017006088.1 4847 Intron XP_016861577.1
XM_017006089.1 4847 UTR 3 XP_016861578.1
XM_017006090.1 4847 UTR 3 XP_016861579.1

View Full Product Details