Product Details
- SNP ID
-
rs147182997
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:49675958 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGCTGGAGAAACATGGGCCTGTTT[A/G]TGAGGATGGTGAGGCATCTGGCTGG
- Phenotype
-
MIM: 102645
MIM: 604020
MIM: 142408
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APEH
PubMed Links
Gene Details
- Gene
- APEH
- Gene Name
- acylaminoacyl-peptide hydrolase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001640.3 |
1734 |
Missense Mutation |
TAT,TGT |
Y145C |
NP_001631.3 |
XM_005265097.2 |
1734 |
Missense Mutation |
TAT,TGT |
Y145C |
XP_005265154.1 |
XM_005265098.4 |
1734 |
Missense Mutation |
TAT,TGT |
Y44C |
XP_005265155.1 |
XM_011533658.2 |
1734 |
Missense Mutation |
TAT,TGT |
Y190C |
XP_011531960.1 |
XM_011533659.1 |
1734 |
Missense Mutation |
TAT,TGT |
Y173C |
XP_011531961.1 |
XM_011533660.2 |
1734 |
Missense Mutation |
TAT,TGT |
Y145C |
XP_011531962.1 |
XM_011533661.2 |
1734 |
Missense Mutation |
ATG,GTG |
M5V |
XP_011531963.1 |
XM_011533662.2 |
1734 |
Missense Mutation |
ATG,GTG |
M5V |
XP_011531964.1 |
XM_011533663.2 |
1734 |
Missense Mutation |
ATG,GTG |
M5V |
XP_011531965.1 |
XM_017006285.1 |
1734 |
Missense Mutation |
TAT,TGT |
Y190C |
XP_016861774.1 |
XM_017006286.1 |
1734 |
Missense Mutation |
TAT,TGT |
Y44C |
XP_016861775.1 |
- Gene
- BSN
- Gene Name
- bassoon presynaptic cytomatrix protein
There are no transcripts associated with this gene.
- Gene
- MST1
- Gene Name
- macrophage stimulating 1
There are no transcripts associated with this gene.
View Full Product Details