Product Details

SNP ID
rs147247805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194686928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCTGTGAGCCTGCACTACTCGGC[A/G]CTCAGCTCGCTGGCGCGGGCGTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM43A PubMed Links

Gene Details

Gene
FAM43A
Gene Name
family with sequence similarity 43 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153690.4 1036 Silent Mutation GCA,GCG A34A NP_710157.2

View Full Product Details