Product Details

SNP ID
rs147528108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49910345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGAGGTCAGTGTCAGGCTCTA[C/G]GTAAGAGATGTGTGCGTGGAAGAAG
Phenotype
MIM: 611464 MIM: 600168
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MON1A PubMed Links

Gene Details

Gene
MON1A
Gene Name
MON1 homolog A, secretory trafficking associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142501.1 1570 Missense Mutation CTA,GTA L320V NP_001135973.1
NM_032355.3 1570 Missense Mutation CTA,GTA L482V NP_115731.2
XM_006713345.3 1570 Missense Mutation CTA,GTA L385V XP_006713408.1
XM_011534160.1 1570 Missense Mutation CTA,GTA L385V XP_011532462.1
Gene
MST1R
Gene Name
macrophage stimulating 1 receptor
There are no transcripts associated with this gene.

View Full Product Details