Product Details

SNP ID
rs148678982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:140678333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAGTTCATCTTCACCTCAGAGC[G/T]GAACTGCACCTGCTTCCCCTGCCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRIM42 PubMed Links

Gene Details

Gene
TRIM42
Gene Name
tripartite motif containing 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152616.4 310 Missense Mutation CAG,CGG Q35R NP_689829.3
XM_011512740.2 310 Missense Mutation CAG,CGG Q35R XP_011511042.1

View Full Product Details