Product Details

SNP ID
rs149045116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:142459273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTCAGAAACCACTCATGAAAAA[C/T]AGGAGGATGCCTGGGCAGGAGAAAT
Phenotype
MIM: 601215
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATR PubMed Links

Gene Details

Gene
ATR
Gene Name
ATR serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184.3 7200 Missense Mutation ATT,GTT I2435V NP_001175.2
XM_011512924.1 7200 Missense Mutation ATT,GTT I2437V XP_011511226.1
XM_011512925.1 7200 Missense Mutation ATT,GTT I2373V XP_011511227.1
XM_011512926.2 7200 Intron XP_011511228.1
XM_017006643.1 7200 Missense Mutation ATT,GTT I2371V XP_016862132.1

View Full Product Details