Product Details

SNP ID
rs149148043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196198302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGAGAGCTGCAGCCTCACCAC[A/G]CCCCGGGGGTCCCTTCCCTGCTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC19 PubMed Links

Gene Details

Gene
ZDHHC19
Gene Name
zinc finger DHHC-type containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039617.1 1015 Missense Mutation GCG,GTG A308V NP_001034706.1
XM_005269279.2 1015 Intron XP_005269336.1
XM_006713493.2 1015 Missense Mutation GCG,GTG A306V XP_006713556.1
XM_006713494.2 1015 Missense Mutation GCG,GTG A282V XP_006713557.1
XM_011512417.1 1015 Missense Mutation GCG,GTG A234V XP_011510719.1

View Full Product Details