Product Details

SNP ID
rs149452125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9964172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATCTAGTGCCCACTGGTGATCC[A/G]TCAGCTCCAAAGCTTCCCACTCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC3 PubMed Links

Gene Details

Gene
EMC3
Gene Name
ER membrane protein complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018447.3 1150 Missense Mutation ACG,ATG T228M NP_060917.1
XM_005265321.3 1150 Intron XP_005265378.1
Gene
PRRT3-AS1
Gene Name
PRRT3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details