Product Details

SNP ID
rs149575792
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:177032335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTGTGAAATGAACCATGGTGGC[A/G]GACACCTACACAAAGAGAAATACAC
Phenotype
MIM: 608628
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TBL1XR1 PubMed Links

Gene Details

Gene
TBL1XR1
Gene Name
transducin (beta)-like 1 X-linked receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321193.1 Intron NP_001308122.1
NM_001321194.1 Intron NP_001308123.1
NM_001321195.1 Intron NP_001308124.1
NM_024665.5 Intron NP_078941.2
XM_005247775.2 Intron XP_005247832.1
XM_006713746.1 Intron XP_006713809.1
XM_011513141.1 Intron XP_011511443.1
XM_011513142.2 Intron XP_011511444.1
XM_011513143.2 Intron XP_011511445.1
XM_017007185.1 Intron XP_016862674.1

View Full Product Details