Product Details
- SNP ID
-
rs150797085
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:9890757 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGCAGGCCCGCGAGCGGCCGGCA[C/T]CGACGGCAGCGACTTTCAGCACCGG
- Phenotype
-
MIM: 616012
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
JAGN1
PubMed Links
Gene Details
- Gene
- JAGN1
- Gene Name
- jagunal homolog 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032492.3 |
171 |
Missense Mutation |
ACC,ATC |
T12I |
NP_115881.3 |
View Full Product Details