Product Details

SNP ID
rs151023095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114295744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTTCAAGGATCGAGTGGCCCC[A/G]GGTCCCGGCCTGGGCCTCACCCTCC
Phenotype
MIM: 612859
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TIGIT PubMed Links

Gene Details

Gene
TIGIT
Gene Name
T-cell immunoreceptor with Ig and ITIM domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173799.3 337 Silent Mutation CCA,CCG P87P NP_776160.2
XM_011512538.1 337 Silent Mutation CCA,CCG P87P XP_011510840.1
XM_017005865.1 337 Intron XP_016861354.1

View Full Product Details