Product Details

SNP ID
rs151173720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69056731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTTCATCTTTATATAATTAAAC[A/C]AAGTCAAAACATAAATTTACATGCA
Phenotype
MIM: 601126 MIM: 603172
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR3136 PubMed Links

Gene Details

Gene
MIR3136
Gene Name
microRNA 3136
There are no transcripts associated with this gene.

Gene
TMF1
Gene Name
TATA element modulatory factor 1
There are no transcripts associated with this gene.

Gene
UBA3
Gene Name
ubiquitin like modifier activating enzyme 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003968.3 Intron NP_003959.3
NM_198195.1 Intron NP_937838.1
XM_011534210.1 Intron XP_011532512.1
XM_011534211.1 Intron XP_011532513.1

View Full Product Details