Product Details

SNP ID
rs151329012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46700203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACTTTACCTCCCACTGTTGAGC[A/G]TTTCCTCGATGGGTCTCCTGGGCTG
Phenotype
MIM: 612402 MIM: 607237
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALS2CL PubMed Links

Gene Details

Gene
ALS2CL
Gene Name
ALS2 C-terminal like
There are no transcripts associated with this gene.

Gene
TMIE
Gene Name
transmembrane inner ear
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147196.2 Intron NP_671729.2
XM_006713097.3 Intron XP_006713160.1

View Full Product Details