Product Details

SNP ID
rs138228830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87340602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCCTCAGGTTCCAGAGTGGGTC[A/C]CAAACTGAAATCAGAGTCAGTCTTC
Phenotype
MIM: 612831
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSD17B11 PubMed Links

Gene Details

Gene
HSD17B11
Gene Name
hydroxysteroid 17-beta dehydrogenase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016245.4 933 Nonsense Mutation GGA,TGA G234* NP_057329.3
XM_011532021.1 933 Intron XP_011530323.1

View Full Product Details