Product Details

SNP ID
rs138288054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1814435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCCAGTGGTTCTAGCTCTTCA[A/C]CTCTGCGACCTCCTTCTCTGCCTTC
Phenotype
MIM: 134934 MIM: 604407
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FGFR3 PubMed Links

Gene Details

Gene
FGFR3
Gene Name
fibroblast growth factor receptor 3
There are no transcripts associated with this gene.

Gene
LETM1
Gene Name
leucine zipper and EF-hand containing transmembrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012318.2 2211 Missense Mutation GTG,TTG V737L NP_036450.1
XM_006713884.1 2211 Missense Mutation GTG,TTG V736L XP_006713947.1
XM_017008187.1 2211 Intron XP_016863676.1

View Full Product Details