Product Details

SNP ID
rs139623858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186588895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGGGATAAAAATTGGGCAAATA[C/T]TGATTCAAGTTGAACCTCTGCCGGT
Phenotype
MIM: 600976
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAT1 PubMed Links

Gene Details

Gene
FAT1
Gene Name
FAT atypical cadherin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005245.3 13652 Silent Mutation CAA,CAG Q4488Q NP_005236.2
XM_005262834.2 13652 Silent Mutation CAA,CAG Q4500Q XP_005262891.1
XM_005262835.2 13652 Silent Mutation CAA,CAG Q4500Q XP_005262892.1
XM_006714139.2 13652 Silent Mutation CAA,CAG Q4488Q XP_006714202.1

View Full Product Details