Product Details

SNP ID
rs140189615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:44678772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGCTACCGACAGGCTCTACAG[A/C]TCCGCAGAATTCAAGGTGACTGCCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GUF1 PubMed Links

Gene Details

Gene
GUF1
Gene Name
GUF1 homolog, GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021927.2 323 Missense Mutation AGA,AGC R50S NP_068746.2
XM_005248122.3 323 UTR 5 XP_005248179.1
XM_011513732.2 323 Missense Mutation AGA,AGC R50S XP_011512034.1

View Full Product Details