Product Details

SNP ID
rs140258264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:24806660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCAAGCCCTCCAGCTGCGGGAG[C/T]AACCTCCCTTGAGGAGAAGTTGTGG
Phenotype
MIM: 185490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC149 PubMed Links

Gene Details

Gene
CCDC149
Gene Name
coiled-coil domain containing 149
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005248210.3 3435 UTR 3 XP_005248267.1
XM_011513906.2 3435 UTR 3 XP_011512208.1
XM_011513907.2 3435 UTR 3 XP_011512209.1
XM_011513908.1 3435 UTR 3 XP_011512210.1
XM_017008827.1 3435 UTR 3 XP_016864316.1
XM_017008828.1 3435 UTR 3 XP_016864317.1
Gene
SOD3
Gene Name
superoxide dismutase 3, extracellular
There are no transcripts associated with this gene.

View Full Product Details